missing translation for 'onlineSavingsMsg'
Learn More

6-Nitro-1,3-dihydroisobenzofuran-1-one, 97%, Thermo Scientific™

Product Code. 10255864
Click to view available options
Quantity:
10 g
50 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 610-93-5
Molecular Formula C8H5NO4
Molecular Weight (g/mol) 179.13
MDL Number MFCD00033529
InChI Key RNWGZXAHUPFXLL-UHFFFAOYSA-N
Synonym 6-nitrophthalide, 6-nitroisobenzofuran-1 3h-one, 6-nitro-3h-isobenzofuran-1-one, 6-nitro-1,3-dihydroisobenzofuran-1-one, 6-nitro-1 3h-isobenzofuranone, 6-nitro-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 6-nitro, 6-nitro-3-hydroisobenzofuran-1-one, 6-nitro-1,3-dihydro-2-benzofuran-1-one, 6-nitrophtalide
PubChem CID 223584
IUPAC Name 6-nitro-1,3-dihydro-2-benzofuran-1-one
SMILES [O-][N+](=O)C1=CC=C2COC(=O)C2=C1

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 179.13
Percent Purity 97%
Chemical Name or Material 6-Nitro-1, 3-dihydroisobenzofuran-1-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'