missing translation for 'onlineSavingsMsg'
Learn More

6-Morpholinopyridine-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10077382
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10077382 1 g Amber Glass Bottle 1g
10113424 250 mg Amber Glass Bottle 250mg
10431334 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10077382 Supplier Thermo Scientific Maybridge Supplier No. CC38301DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 554405-17-3
Molecular Formula C10H12N2O3
Molecular Weight (g/mol) 208.217
InChI Key SAZSDHZXRMXESN-UHFFFAOYSA-N
Synonym 6-morpholinopyridine-2-carboxylic acid, 6-morpholinopicolinic acid, 6-morpholin-4-yl pyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 6-4-morpholinyl, 6-morpholinopyridine-2-carboxylicacid, 6-morpholin-4-yl picolinic acid, 6-4-morpholinyl-2-pyridinecarboxylic acid
PubChem CID 2388921
IUPAC Name 6-morpholin-4-ylpyridine-2-carboxylic acid
SMILES C1COCCN1C2=CC=CC(=N2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 208.22
Percent Purity 97%
Chemical Name or Material 6-Morpholinopyridine-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.