missing translation for 'onlineSavingsMsg'
Learn More

(6-morpholinopyrid-2-yl)methylamine, 95%, Thermo Scientific™

Product Code. 10608303
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10608303 250 mg Amber Glass Bottle 250mg
10620654 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10608303 Supplier Thermo Scientific Maybridge Supplier No. CC38313CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 868755-52-6
Molecular Formula C10H15N3O
Molecular Weight (g/mol) 193.25
MDL Number MFCD08435862
InChI Key JRKWUCQBVPZBBP-UHFFFAOYSA-N
Synonym 6-morpholinopyrid-2-yl methylamine, 6-morpholin-4-yl pyridin-2-yl methanamine, 2-pyridinemethanamine,6-4-morpholinyl, 6-morpholin-4-ylpyridin-2-yl methylamine, 1-6-morpholin-4-yl pyridin-2-yl methanamine, 6-morpholinopyridin-2-yl methanamine, 6-morpholinopyridin-2-yl methylamine, 6-morpholin-4-yl-2-pyridyl methylamine, 2-aminomethyl-6-morpholin-4-yl pyridine
PubChem CID 18525737
SMILES NCC1=NC(=CC=C1)N1CCOCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 1-[6-(morpholin-4-yl)pyridin-2-yl]methanamine
Formula Weight 193.25
Percent Purity 95%
Chemical Name or Material (6-Morpholinopyrid-2-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.