missing translation for 'onlineSavingsMsg'
Learn More

6-Morpholino-3-pyridinyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10089221
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10089221 1 g 1g
10089211 250 mg 250mg
10284524 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10089221 Supplier Thermo Scientific Maybridge Supplier No. CC17507DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 52024-29-0
Molecular Formula C10H11N3OS
Molecular Weight (g/mol) 221.278
MDL Number MFCD03086114
InChI Key VLZWMPRZGPULIU-UHFFFAOYSA-N
Synonym 4-5-isothiocyanatopyridin-2-yl morpholine, 6-morpholino-3-pyridinyl isothiocyanate, 6-morpholino-3-pyridinylisothiocyanate, 4-5-isothiocyanato-2-pyridyl morpholine, 4-5-isothiocyatopyridin-2-yl morpholine, 6-morpholin-4-ylpyridin-3-isothiocyanate, 4-5-isothiocyanato-2-pyridinyl morpholine, 5-isothiocyanato-2-morpholin-4-yl pyridine, morpholine,4-5-isothiocyanato-2-pyridinyl, 6-morpholin-4-yl pyridin-3-yl isothiocyanate
PubChem CID 2776464
IUPAC Name 4-(5-isothiocyanatopyridin-2-yl)morpholine
SMILES C1COCCN1C2=NC=C(C=C2)N=C=S

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 221.28
Percent Purity 97%
Chemical Name or Material 6-Morpholino-3-pyridinyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.