missing translation for 'onlineSavingsMsg'
Learn More

6-Morpholin-4-yl-pyridine-3-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. p-2703423
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10477612

Brand: Thermo Scientific Maybridge CC17503CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 337508-68-6
Molecular Formula C9H11ClN2O3S
Molecular Weight (g/mol) 262.708
MDL Number MFCD02681929
InChI Key FZQDEGBLWSULKG-UHFFFAOYSA-N
Synonym 6-morpholin-4-yl-pyridine-3-sulfonyl chloride, 6-morpholinopyridine-3-sulfonyl chloride, 6-morpholin-4-yl pyridine-3-sulfonyl chloride, 3-chlorosulphonyl-6-morpholin-4-yl pyridine, 3-pyridinesulfonyl chloride, 6-4-morpholinyl, 6-morpholin-4-yl pyridine-3-sulphonyl chloride, chloro 6-morpholin-4-yl 3-pyridyl sulfone, 6-4-morpholinyl-3-pyridinesulfonyl chloride, 3-pyridinesulfonylchloride, 6-4-morpholinyl, 4-5-chlorosulfonyl pyridin-2-yl morpholine
PubChem CID 2776462
IUPAC Name 6-morpholin-4-ylpyridine-3-sulfonyl chloride
SMILES C1COCCN1C2=NC=C(C=C2)S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 262.72
Percent Purity 97%
Chemical Name or Material 6-Morpholin-4-yl-pyridine-3-sulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.