missing translation for 'onlineSavingsMsg'
Learn More

6-Methyluracil, 97%

Product Code. 11491227
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491227 2500 g 2500g
11471227 100 g 100g
11481227 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491227 Supplier Thermo Scientific Alfa Aesar Supplier No. B24191.0E
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 626-48-2 | C5H6N2O2 | 126.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-48-2
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00006028
InChI Key SHVCSCWHWMSGTE-UHFFFAOYSA-N
Synonym 6-methyluracil, 2,4-dihydroxy-6-methylpyrimidine, pseudothymine, 6-methylpyrimidine-2,4 1h,3h-dione, 4-methyluracil, 6-methylpyrimidine-2,4-diol, 2,4 1h,3h-pyrimidinedione, 6-methyl, uracil, 6-methyl, 6-methyl-2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 6-methyl
PubChem CID 12283
ChEBI CHEBI:74733
IUPAC Name 6-methyl-1H-pyrimidine-2,4-dione
SMILES CC1=CC(=O)NC(=O)N1

Specifications

Melting Point 317°C to 320°C
Quantity 2500 g
Beilstein 115647
Merck Index 14,6133
Formula Weight 126.12
Percent Purity 97%
Chemical Name or Material 6-Methyluracil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.