missing translation for 'onlineSavingsMsg'
Learn More

6-(Methylthio)pyridine-3-boronic acid, 95%, Thermo Scientific™

Product Code. 15493797
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493797 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15493797 Supplier Thermo Scientific Alfa Aesar Supplier No. H27873.MD
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 321438-86-2
Molecular Formula C6H8BNO2S
MDL Number MFCD03788238
Quantity 250 mg
Synonym 2-methylthiopyridine-5-boronic acid, 6-methylthio pyridin-3-ylboronic acid, 2-methylthio-5-pyridinyl-boronic acid, 2-methylthio-pyridine-5-boronic acid, 6-methylthio pyridin-3-yl boronic acid, 6-methylsulphanyl pyridine-3-boronic acid, 6-methylthio pyridin-3-yl-3-boronic acid, 6-methylthio pyridine-3-boronic acid, 5-borono-2-methylmercapto pyridine, 6-methylsulfanyl pyridin-3-ylboronic acid
InChI Key UXWKVPJOPVIIRU-UHFFFAOYSA-N
SMILES B(C1=CN=C(C=C1)SC)(O)O
IUPAC Name (6-methylsulfanylpyridin-3-yl)boronic acid
Molecular Weight (g/mol) 169.005
PubChem CID 2762705
Formula Weight 169.01
Percent Purity 95%
Chemical Name or Material 6-(Methylthio)pyridine-3-boronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.