missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 6-(Methylthio)purine, 97%
Click to view available options
Quantity:
1g
Unit Size:
1g
Description
Applications
6-(Methylthio)purine can react with 1-bromo-2-chloro-ethane to produce 6-methylsulfanyl-9-vinyl-9H-purine. This reaction will need reagent NaOH, catalyst (C8H17)4NBr, and the menstruum benzene and H2O.
Solubility
Soluble in water (769 mg/L).
Notes
Keep container tightly closed. Store away from oxidizing agents.
6-(Methylthio)purine can react with 1-bromo-2-chloro-ethane to produce 6-methylsulfanyl-9-vinyl-9H-purine. This reaction will need reagent NaOH, catalyst (C8H17)4NBr, and the menstruum benzene and H2O.
Solubility
Soluble in water (769 mg/L).
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
| CAS | 50-66-8 |
| Molecular Formula | C6H6N4S |
| MDL Number | MFCD00005576 |
| Quantity | 1g |
| Beilstein | 7695 |
| Synonym | 6-methylthio purine, 6-methylmercaptopurine, 6-methylthiopurine, 1h-purine, 6-methylthio, 6-methyl mp, purine, 6-methylthio, s-methyl-6-mercaptopurine, 6-methylsulfanyl-9h-purine, unii-6v404dv25o, thiopurine s-methylether |
| InChI Key | UIJIQXGRFSPYQW-UHFFFAOYSA-N |
| SMILES | CSC1=C2NC=NC2=NC=N1 |
| IUPAC Name | 6-methylsulfanyl-7H-purine |
| Molecular Weight (g/mol) | 166.20 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction