missing translation for 'onlineSavingsMsg'
Learn More

6-Methylpyridine-3-carboxylic Acid (6-Methylnicotinic Acid), Mikromol™

Product Code. 17971726
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971726 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17971726 Supplier Mikromol Supplier No. MM0553.050025
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

6-Methylpyridine-3-carboxylic Acid (6-Methylnicotinic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Methylnicotinic Acid
CAS 3222-47-7
Analyte or Component Names 6-Methylnicotinic Acid
Shipping Condition Room Temperature
Molecular Formula C7 H7 N O2
Certifications/Compliance ISO 17025
Synonym 6-Methylpyridine-3-carboxylic Acid, 6-Methylnicotinic Acid, 3-Pyridinecarboxylic acid, 6-methyl-, Nicotinic acid, 6-methyl- (6CI,7CI,8CI), 6-Methyl-3-pyridinecarboxylic acid, 2-Methyl-5-pyridinecarboxylic acid, 2-Picoline-5-carboxylic acid, 5-Carboxy-2-methylpyridine, 6-Methylnicotinic acid, NSC 135040, NSC 527351, Nicotinic Acid Impurity A
InChI Formula InChI=1S/C7H7NO2/c1-5-2-3-6(4-8-5)7(9)10/h2-4H,1H3,(H,9,10)
SMILES Cc1ccc(cn1)C(=O)O
IUPAC Name 6-methylpyridine-3-carboxylic acid
Molecular Weight (g/mol) 137.14
Quantity 25 mg
Formula Weight 137.0477 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.