missing translation for 'onlineSavingsMsg'
Learn More

6-Methylpyridine-2-carboxaldehyde, 98%

Product Code. 11326999
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326999 25 g 25g
11316999 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326999 Supplier Thermo Scientific Alfa Aesar Supplier No. H26841.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1122-72-1 | C7H7NO | 121.139 g/mol

6-Methyl-2-pyridinecarboxaldehyde was used in the synthesis of 1-(10-cyano- and -10-bromo-5H-dibenzo[a,d]cyclohepten-5-yl)-4-[(arylmethylene)amino]piperazines which are compounds that exhibits anticon vulsant properties. 6-Methylpyridine-2-carboxaldehyde is used to produce 1-(6-methyl-pyridin-2-yl)-hexan-1-one with pent-1-ene at temperature of 100°C. This reaction will need reagent 2-amino-3-picoline and catalysts (Ph3P)3RhCl, Cp2ZrCl2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-2-pyridinecarboxaldehyde was used in the synthesis of 1-(10-cyano- and -10-bromo-5H-dibenzo[a,d]cyclohepten-5-yl)-4-[(arylmethylene)amino]piperazines which are compounds that exhibits anticon vulsant properties. 6-Methylpyridine-2-carboxaldehyde is used to produce 1-(6-methyl-pyridin-2-yl)-hexan-1-one with pent-1-ene at temperature of 100°C. This reaction will need reagent 2-amino-3-picoline and catalysts (Ph3P)3RhCl, Cp2ZrCl2.

Solubility
Soluble in water (750 g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-72-1
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00006291
InChI Key AHISYUZBWDSPQL-UHFFFAOYSA-N
Synonym 6-methyl-2-pyridinecarboxaldehyde, 6-methylpicolinaldehyde, 2-formyl-6-methylpyridine, 6-methylpyridine-2-carboxaldehyde, 2-pyridinecarboxaldehyde, 6-methyl, 6-methylpyridine-2-aldehyde, 2-formyl-6-picoline, 6-methyl-2-pyridinecarbaldehyde, 6-methyl-2-picolinaldehyde, 6-methylpyridyl-2-aldehyde
PubChem CID 70737
IUPAC Name 6-methylpyridine-2-carbaldehyde
SMILES CC1=CC=CC(=N1)C=O

Specifications

Melting Point 29°C to 33°C
Boiling Point 105°C to 108°C (12 mmHg)
Flash Point 68°C (155°F)
Refractive Index 1.527
Quantity 25 g
Beilstein 107476
Solubility Information Soluble in water (750g/L).
Formula Weight 121.14
Percent Purity 98%
Chemical Name or Material 6-Methylpyridine-2-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.