missing translation for 'onlineSavingsMsg'
Learn More

6-Methylpicolinic acid, 97%, ACROS Organics™

Product Code. 10061414 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10061414 1 g Glass bottle 1g
10163154 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10061414 Supplier Thermo Scientific Acros Supplier No. 425660010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 934-60-1
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00023481
InChI Key LTUUGSGSUZRPRV-UHFFFAOYSA-N
Synonym 6-methyl-2-pyridinecarboxylic acid, 6-methylpicolinic acid, 6-methyl-2-picolinic acid, 2-carboxy-6-methylpyridine, 6-methyl-2-picolinicacid, 2-pyridinecarboxylic acid, 6-methyl, 2-picoline-6-carboxylic acid, picolinic acid, 6-methyl, 6-methyl picolinic acid, unii-kb675y352l
PubChem CID 70282
IUPAC Name 6-methylpyridine-2-carboxylic acid
SMILES CC1=CC=CC(=N1)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 120.0°C to 127.0°C
Boiling Point 100.0°C (4.5 mmHg)
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 137.14
Percent Purity 97%
Chemical Name or Material 6-Methylpicolinic acid
compliance-icons
Product Identifier
  • 6-Methylpicolinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.