missing translation for 'onlineSavingsMsg'
Learn More

6-Methylnicotinic acid, 99%

Product Code. 11352766
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352766 25 g 25g
11332766 1 g 1g
11342766 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11352766 Supplier Thermo Scientific Alfa Aesar Supplier No. L08877.14
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3222-47-7 | C7H7NO2 | 137.138 g/mol

6-Methylnicotinic acid is an intermediate of the drug etoricoxib (a non- steroidal anti-inflammatory drug for the treatment of arthritis and osteoarthritis). It is also used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methylnicotinic acid is an intermediate of the drug etoricoxib (a non- steroidal anti-inflammatory drug for the treatment of arthritis and osteoarthritis). It is also used as organic intermediates.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3222-47-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00006341
InChI Key RZOKQIPOABEQAM-UHFFFAOYSA-N
Synonym 6-methylnicotinic acid, 3-pyridinecarboxylic acid, 6-methyl, 6-methyl nicotinic acid, 2-methylpyridine-5-carboxylic acid, 2-methyl-5-pyridinecarboxylic acid, 6-methyl-3-pyridinecarboxylic acid, 2-methyl-5-pyridine carboxylic acid, 6-methylnicotinate, 6-methylnicotinicacid, pubchem1218
PubChem CID 137860
IUPAC Name 6-methylpyridine-3-carboxylic acid
SMILES CC1=NC=C(C=C1)C(=O)O

Specifications

Melting Point 210°C to 215°C
Quantity 25 g
Beilstein 112049
Solubility Information Sparingly soluble in water.
Formula Weight 137.14
Percent Purity 99%
Chemical Name or Material 6-Methylnicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.