missing translation for 'onlineSavingsMsg'
Learn More

6-Methylimidazo[2,1-b][1,3]thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10432474
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10432474 10 g Amber Glass Bottle 10g
10120932 250 mg Amber Glass Bottle 250mg
10772113 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10432474 Supplier Thermo Scientific Maybridge Supplier No. MO00015EA
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 77628-51-4
Molecular Formula C7H6N2O2S
Molecular Weight (g/mol) 182.20
MDL Number MFCD02083132
InChI Key VSQMBZXCDDTZAF-UHFFFAOYSA-N
Synonym 6-methylimidazo 2,1-b 1,3 thiazole-5-carboxylic acid, 6-methylimidazo 2,1-b thiazole-5-carboxylic acid, 6-methylimidazo 2,1-b 1,3-thiazoline-5-carboxylic acid, 6-methyl-imidazo 2,1-b thiazole-5-carboxylic acid, 6-methyl-imidazo 2.1-b thiazole-5-carboxylic acid, imidazo 2,1-b thiazole-5-carboxylic acid, 6-methyl, imidazo 2,1-b thiazole-5-carboxylicacid, 6-methyl, 6-methylimidazo 2,3-b 1,3 thiazole-5-carboxylic acid, 6-methylimidazo 2,1-b 1,3 thiazole-5-carboxylic a
PubChem CID 329979
IUPAC Name 6-methylimidazo[2,1-b][1,3]thiazole-5-carboxylic acid
SMILES CC1=C(N2C=CSC2=N1)C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 182.2
Percent Purity 97%
Chemical Name or Material 6-Methylimidazo[2, 1-b][1, 3]thiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.