missing translation for 'onlineSavingsMsg'
Learn More

6-Methylheptanol (90%), TRC

Product Code. 30480263
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480263 2.5 g 2.5g
30464409 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30480263 Supplier Toronto Research Chemicals Supplier No. TRCM3119302.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1653-40-3
Molecular Formula C8 H18 O
Molecular Weight (g/mol) 130.23
Synonym 1-Heptanol, 6-methyl- (6CI, 8CI, 9CI, ACI), 6-Methyl-1-heptanol (ACI), 6-Methylheptanol, 6-Methyl-1-heptyl alcohol
IUPAC Name 6-methylheptan-1-ol
SMILES CC(C)CCCCCO

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C8H18O/c1-8(2)6-4-3-5-7-9/h8-9H,3-7H2,1-2H3
Formula Weight 130.1358
Chemical Name or Material 6-Methylheptanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.