missing translation for 'onlineSavingsMsg'
Learn More

6-Methylheptane-2,4-dione, 98+%

Product Code. 11491003
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491003 10 g 10g
11374639 2 g 2g
11401013 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491003 Supplier Thermo Scientific Alfa Aesar Supplier No. A13373.09
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3002-23-1 | C8H14O2 | 142.198 g/mol

6-Methylheptane-2,4-dione, is used as a multipurpose intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceutical industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methylheptane-2,4-dione, is used as a multipurpose intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceutical industry

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3002-23-1
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00008939
InChI Key IGMOYJSFRIASIE-UHFFFAOYSA-N
Synonym 6-methyl-2,4-heptanedione, isovalerylacetone, 2,4-heptanedione, 6-methyl, unii-o97vug08ub, 6-methylheptan-2,4-dione, o97vug08ub, nsc94, nsc 94, acmc-1ct2p, 6-methyl-2,4-heptandione
PubChem CID 76354
IUPAC Name 6-methylheptane-2,4-dione
SMILES CC(C)CC(=O)CC(=O)C

Specifications

Density 0.919
Boiling Point 183°C
Flash Point 58°C (136°F)
Refractive Index 1.456
Quantity 10 g
UN Number UN1224
Beilstein 1751122
Solubility Information It is insoluble in water.
Formula Weight 142.2
Percent Purity ≥98%
Chemical Name or Material 6-Methylheptane-2,4-dione
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.