missing translation for 'onlineSavingsMsg'
Learn More

6-Methylchromone hydrate, 99%, Thermo Scientific™

Product Code. 10281033 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10281033 1 g Glass Bottle 1g
10545833 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10281033 Supplier Thermo Scientific Acros Supplier No. 373020010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 207511-19-1
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00209598
InChI Key HTXQVFXXVXOLCF-UHFFFAOYSA-N
Synonym 6-methylchromone hydrate, 6-methyl-4h-chromen-4-one hydrate, 6-methylchromen-4-one hydrate, 6-methyl-4h-1-benzopyran-4-one-water 1/1
PubChem CID 16212708
SMILES CC1=CC=C2OC=CC(=O)C2=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 70°C to 75°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 1 g
IUPAC Name 6-methyl-4H-chromen-4-one
Formula Weight 160.17
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 6-Methylchromone hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.