missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl Nicotinic Acid, TRC

Product Code. 30483612
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483612 1 g 1g
30480304 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30483612 Supplier Toronto Research Chemicals Supplier No. TRCM3233301G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3222-47-7
Molecular Formula C7 H7 N O2
Molecular Weight (g/mol) 137.14
Synonym 6-Methylpyridine-3-carboxylic Acid, 6-Methylnicotinic Acid, 3-Pyridinecarboxylic acid, 6-methyl-, Nicotinic acid, 6-methyl- (6CI,7CI,8CI), 6-Methyl-3-pyridinecarboxylic acid, 2-Methyl-5-pyridinecarboxylic acid, 2-Picoline-5-carboxylic acid, 5-Carboxy-2-methylpyridine, 6-Methylnicotinic acid, NSC 135040, NSC 527351
IUPAC Name 6-methylpyridine-3-carboxylic acid
SMILES Cc1ccc(cn1)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H7NO2/c1-5-2-3-6(4-8-5)7(9)10/h2-4H,1H3,(H,9,10)
Formula Weight 137.0477
Chemical Name or Material 6-Methylnicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.