missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl Imatinib, TRC

Product Code. 30467825
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467825 10 mg 10mg
30483260 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30467825 Supplier Toronto Research Chemicals Supplier No. TRCM21434010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Methyl Imatinib
CAS 1032314-85-4
Molecular Formula C30 H33 N7 O
Quantity 10 mg
Synonym N-[4-Methyl-3-[[4-(6-methyl-3-pyridinyl)-2-pyrimidinyl]amino]phenyl]-4-[(4-methyl-1-piperazinyl)methyl]benzamide, N-[5-[4-[(4-Methylpiperazino)methyl]benzoylamino]-2-methylphenyl]-4-(6-methylpyridin-3-yl)-2-pyrimidineamine, N-(4-Methyl-3-((4-(6-methylpyridin-3-yl)pyrimidin-2-yl)amino)phenyl)-4-((4-methylpiperazin-1-yl)methyl)benzamide
InChI Formula InChI=1S/C30H33N7O/c1-21-4-11-26(18-28(21)35-30-31-13-12-27(34-30)25-8-5-22(2)32-19-25)33-29(38)24-9-6-23(7-10-24)20-37-16-14-36(3)15-17-37/h4-13,18-19H,14-17,20H2,1-3H3,(H,33,38)(H,31,34,35)
SMILES CN1CCN(Cc2ccc(cc2)C(=O)Nc3ccc(C)c(Nc4nccc(n4)c5ccc(C)nc5)c3)CC1
IUPAC Name N-[4-methyl-3-[[4-(6-methylpyridin-3-yl)pyrimidin-2-yl]amino]phenyl]-4-[(4-methylpiperazin-1-yl)methyl]benzamide
Molecular Weight (g/mol) 507.629
Formula Weight 507.275
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.