missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-5-hepten-2-one, 98%

Product Code. 11450953
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11450953

Brand: Thermo Scientific Alfa Aesar A13335.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-93-0 | C8H14O | 126.20 g/mol

6-Methyl-hepten-2-one is used in the synthesis of thyrsiferyl 23-Acetate which acts as an anti-leukemic inducer of apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-hepten-2-one is used in the synthesis of thyrsiferyl 23-Acetate which acts as an anti-leukemic inducer of apoptosis.

Solubility
Soluble in methanol and chloroform.

Notes
Store in cool, dry place in tightly closed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-93-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00008905
InChI Key UHEPJGULSIKKTP-UHFFFAOYSA-N
Synonym 6-methyl-5-hepten-2-one, sulcatone, methylheptenone, 5-hepten-2-one, 6-methyl, 2-methyl-2-hepten-6-one, methyl heptenone, 6-methyl-5-heptene-2-one, heptenone, methyl, 2-methyl-6-oxo-2-heptene, 2-oxo-6-methylhept-5-ene
PubChem CID 9862
ChEBI CHEBI:16310
IUPAC Name 6-methylhept-5-en-2-one
SMILES CC(C)=CCCC(C)=O

Specifications

Melting Point -67°C
Density 0.855
Boiling Point 172°C to 174°C
Flash Point 56°C (133°F)
Refractive Index 1.439
Quantity 100 g
UN Number UN1224
Beilstein 1741705
Solubility Information Soluble in methanol and chloroform.
Formula Weight 126.2
Percent Purity 98%
Chemical Name or Material 6-Methyl-5-hepten-2-one
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.