missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-4-trifluoromethyl-2(1H)-pyridone, 97%

Product Code. 11345787
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
25 g
Unit Size:
0.25g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11345787 25 g 25g
11420590 0.25 g 0.25g
11335787 1 g 1g
11325787 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11345787 Supplier Thermo Scientific Alfa Aesar Supplier No. 043467.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 22123-19-9 | C7H6F3NO | 177.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22123-19-9
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD00042479
InChI Key CDRMKCNOGIBXOG-UHFFFAOYSA-N
Synonym 6-methyl-4-trifluoromethyl pyridin-2 1h-one, 6-methyl-4-trifluoromethyl-2 1h-pyridinone, 6-methyl-4-trifluoromethyl pyridin-2-ol, 6-methyl-4-trifluoromethyl-1h-pyridin-2-one, 6-methyl-4-trifluoromethyl-2 1h-pyridone, 2 1h-pyridinone,6-methyl-4-trifluoromethyl, 2-hydroxy-6-methyl-4-trifluoromethyl pyridine, 6-methyl-4-trifluoromethyl-1,2-dihydropyridin-2-one, 6-methyl-4-trifluoromethyl hydropyridin-2-one, acmc-1cb5l
PubChem CID 2782992
IUPAC Name 6-methyl-4-(trifluoromethyl)-1H-pyridin-2-one
SMILES CC1=CC(=CC(=O)N1)C(F)(F)F

Specifications

Melting Point 129°C to 130°C
Quantity 25 g
Formula Weight 177.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Methyl-4-trifluoromethyl-2 (1H)-pyridone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.