missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-3-pyridoyl-2-pyrrolidinone, TRC

Product Code. 30477526
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30477526 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477526 Supplier Toronto Research Chemicals Supplier No. TRCM325944250MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Methyl-3-pyridoyl-2-pyrrolidinone
CAS 1076198-57-6
Molecular Formula C11 H12 N2 O2
Quantity 250 mg
Synonym 2-Pyrrolidinone, 3-[(6-methyl-3-pyridinyl)carbonyl]-, 6-Methyl-3-pyridoyl-2-pyrrolidinone, 1-(6-Methylnicotinoyl)pyrrolidin-2-one, 3-(6-Methylnicotinoyl)pyrrolidin-2-one, 3-[(6-Methyl-3-pyridinyl)carbonyl]-2-pyrrolidinone
InChI Formula InChI=1S/C11H12N2O2/c1-8-4-5-9(7-12-8)11(15)13-6-2-3-10(13)14/h4-5,7H,2-3,6H2,1H3
SMILES Cc1ccc(cn1)C(=O)N2CCCC2=O
IUPAC Name 1-(6-methylpyridine-3-carbonyl)pyrrolidin-2-one
Molecular Weight (g/mol) 204.23
Formula Weight 204.0899
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.