missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-3,5-heptadien-2-one, 97%

Product Code. 11387256
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387256 25 g 25g
11377256 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387256 Supplier Thermo Scientific Alfa Aesar Supplier No. L09169.14
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1604-28-0 | C8H12O | 124.183 g/mol

6-Methyl-3,5-heptadien-2-one is used in the preparation of (RS)-(E)-2,6-dimethyl-1,2-epoxy-3,5-heptadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-3,5-heptadien-2-one is used in the preparation of (RS)-(E)-2,6-dimethyl-1,2-epoxy-3,5-heptadiene.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1604-28-0
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
MDL Number MFCD00043647
InChI Key KSKXSFZGARKWOW-GQCTYLIASA-N
Synonym 6-methyl-3,5-heptadien-2-one, 6-methylhepta-3,5-dien-2-one, methylheptadienone, unii-p7cmp2e76c, 3,5-heptadien-2-one, 6-methyl, 3e-6-methylhepta-3,5-dien-2-one, 3,5-heptadien-2-one, 6-methyl-, e, 6-methyl-trans-3,5-heptadien-2-one, p7cmp2e76c, fema no. 3363
PubChem CID 5370101
IUPAC Name (3E)-6-methylhepta-3,5-dien-2-one
SMILES CC(=CC=CC(=O)C)C

Specifications

Density 0.89
Boiling Point 95°C to 96°C (20 mmHg)
Odor Pleasant
Refractive Index 1.534
Quantity 25 g
Beilstein 1699937
Solubility Information Miscible with alcohol. Immiscible with water.
Formula Weight 124.18
Percent Purity 97%
Chemical Name or Material 6-Methyl-3,5-heptadien-2-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.