missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-2-pyridinemethanol, 98%

Product Code. 11326899
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326899 25 g 25g
11306899 1 g 1g
11316899 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326899 Supplier Thermo Scientific Alfa Aesar Supplier No. H26794.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1122-71-0 | C7H9NO | 123.16 g/mol

Use as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use as primary and secondary intermediate.

Solubility
Low soluble in water, high soluble in ethanol, acetic acid. Soluble in benzene, toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, strong reducing agents. Store under an inert atmosphere.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-71-0
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00023520
InChI Key JLVBSBMJQUMAMW-UHFFFAOYSA-N
Synonym 6-methyl-2-pyridinemethanol, 6-methylpyridin-2-yl methanol, 2-pyridinemethanol, 6-methyl, 6-methyl-2-pyridylmethanol, 6-methylpyridine-2-methanol, 2-hydroxymethyl-6-methylpyridine, 6-methyl-2-pyridyl methan-1-ol, pubchem20959, acmc-2099dw, 6-methyl-2-pyridylcarbinol
PubChem CID 70736
IUPAC Name (6-methylpyridin-2-yl)methanol
SMILES CC1=CC=CC(CO)=N1

Specifications

Melting Point 32°C to 34°C
Boiling Point 105°C to 108°C (12 mmHg)
Flash Point 109°C (228°F)
Quantity 25 g
Solubility Information Low soluble in water,high soluble in ethanol,acetic acid. Soluble in benzene,toluene.
Formula Weight 123.15
Percent Purity 98%
Chemical Name or Material 6-Methyl-2-pyridinemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.