missing translation for 'onlineSavingsMsg'
Learn More

6-methyl-2,3-dihydropyridazin-3-one hydrate, Thermo Scientific™

Product Code. 10492483
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10492483 10 g 10g
10537893 1 g 1g
10652265 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10492483 Supplier Thermo Scientific Maybridge Supplier No. BTB06840EA
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 7143-82-0
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00039720
InChI Key QZWIXLPWMGHDDD-UHFFFAOYSA-N
Synonym 6-methylpyridazin-3-ol hydrate, 6-methylpyridazin-3 2h-one hydrate, 6-methyl-2,3-dihydropyridazin-3-one hydrate, 6-methyl-2h-pyridazin-3-one hydrate, 6-methylpyridazin-3-one hydrate, 3-methyl-1h-pyridazin-6-one hydrate, 6-methylpyridazin-3 2h ;-one hydrate, 6-methyl-2-hydropyridazin-3-one, hydrate, 6-methyl-2,3-dihydropyridazine-3-one hydrate, 6-methylpyridazin-3 2h-one-water 1/1
PubChem CID 2774632
SMILES CC1=NNC(=O)C=C1

Specifications

Assay Percent Range 90%
Quantity 10 g
IUPAC Name 3-methyl-1H-pyridazin-6-one;hydrate
Formula Weight 128.14
Percent Purity 90%
Chemical Name or Material 6-Methyl-2, 3-dihydropyridazin-3-one hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.