missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-1,2,3,4-tetrahydroquinoline, 98%

Product Code. 11452298
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452298 50 g 50g
11442298 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11452298 Supplier Thermo Scientific Alfa Aesar Supplier No. A15245.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-61-2 | C10H13N | 147.22 g/mol

6-Methyl-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-61-2
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.22
MDL Number MFCD00023887
InChI Key XOKMRXSMOHCNIX-UHFFFAOYSA-N
Synonym civettal, 1,2,3,4-tetrahydro-6-methylquinoline, quinoline, 1,2,3,4-tetrahydro-6-methyl, quinoline, tetrahydro-6-methyl, p-methyltetrahydroquinoline, acmc-209rck, dsstox_cid_27444, dsstox_rid_82351, dsstox_gsid_47444, xokmrxsmohcnix-uhfffaoysa
PubChem CID 66678
IUPAC Name 6-methyl-1,2,3,4-tetrahydroquinoline
SMILES CC1=CC=C2NCCCC2=C1

Specifications

Melting Point 33°C to 37°C
Boiling Point 262°C to 263°C (712 mmHg)
Quantity 50 g
Beilstein 122515
Solubility Information Insoluble in water.
Formula Weight 147.22
Percent Purity 98%
Chemical Name or Material 6-Methyl-1,2,3,4-tetrahydroquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.