missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxytryptamine, 99%

Product Code. 10237303 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
50mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10237303 50 mg Glass bottle 50mg
10327800 250 mg Glass bottle 250mg
10165583 1 g Glass bottle 1g
11906621 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10237303 Supplier Thermo Scientific Acros Supplier No. 187450500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3610-36-4 | C11H14N2O | 190.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3610-36-4
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.24
MDL Number MFCD00005663
InChI Key VOCGEKMEZOPDFP-UHFFFAOYSA-N
PubChem CID 17654
IUPAC Name 2-(6-methoxy-1H-indol-3-yl)ethanamine
SMILES COC1=CC2=C(C=C1)C(=CN2)CCN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 143.0°C to 146.0°C
Assay Percent Range 99%
Packaging Glass bottle
Quantity 50 mg
Synonym 6-methoxytryptamine, 2-6-methoxy-1h-indol-3-yl ethanamine, 3-2-aminoethyl-6-methoxyindole, 2-6-methoxy-1h-indol-3-yl-ethylamine, 1h-indole-3-ethanamine, 6-methoxy, 2-2-aminoethyl-5-methoxyindole, 1h-indole-3-ethanamine,6-methoxy, 2-6-methoxy-1h-indol-3-yl ethan-1-amine, 2-6-methoxyindol-3-yl ethylamine, 6-methoxy tryptamine
Solubility Information Solubility in water: moderately soluble. Other solubilities: soluble in ethanol
Formula Weight 190.24
Percent Purity 99%
Chemical Name or Material 6-Methoxytryptamine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 6-Methoxytryptamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.