Learn More
6-Methoxyquinoline, 98%, Thermo Scientific Chemicals
£50.90 - £166.00
Chemical Identifiers
CAS | 5263-87-6 |
---|---|
Molecular Formula | C10H9NO |
Molecular Weight (g/mol) | 159.19 |
MDL Number | MFCD00006800 |
InChI Key | HFDLDPJYCIEXJP-UHFFFAOYSA-N |
Synonym | quinoline, 6-methoxy, 6-methoxy-quinoline, methyl 6-quinolyl ether, p-quinanisole, unii-5s1u3125ad, chembl15200, 6-methoxy quinoline, pubchem5917, 6-methoxyquinoline, acmc-209l0q |
PubChem CID | 14860 |
ChEBI | CHEBI:72822 |
IUPAC Name | 6-methoxyquinoline |
SMILES | COC1=CC2=C(C=C1)N=CC=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10052280
|
Thermo Scientific Acros
179340250 |
25 g | Glass bottle |
£50.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13428428
|
Thermo Scientific Acros
179341000 |
100 g | Glass bottle |
£166.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5263-87-6 | |
159.19 | |
HFDLDPJYCIEXJP-UHFFFAOYSA-N | |
14860 | |
6-methoxyquinoline |
C10H9NO | |
MFCD00006800 | |
quinoline, 6-methoxy, 6-methoxy-quinoline, methyl 6-quinolyl ether, p-quinanisole, unii-5s1u3125ad, chembl15200, 6-methoxy quinoline, pubchem5917, 6-methoxyquinoline, acmc-209l0q | |
CHEBI:72822 | |
COC1=CC2=C(C=C1)N=CC=C2 |
Specifications
5263-87-6 | |
100.0 | |
White to Cream | |
193.0°C (50.0 mmHg) | |
Authentic | |
C10H9NO | |
MFCD00006800 | |
1.152 | |
Solubility in water: insoluble | |
COC1=CC2=C(C=C1)N=CC=C2 | |
159.19 | |
CHEBI:72822 | |
98% | |
6-Methoxyquinoline, 98% |
97.5 | |
18.0°C to 20.0°C | |
1.1520g/mL | |
>110°C | |
97.5% min. (GC) | |
1.6170 to 1.6240 | |
21,85 | |
quinoline, 6-methoxy, 6-methoxy-quinoline, methyl 6-quinolyl ether, p-quinanisole, unii-5s1u3125ad, chembl15200, 6-methoxy quinoline, pubchem5917, 6-methoxyquinoline, acmc-209l0q | |
HFDLDPJYCIEXJP-UHFFFAOYSA-N | |
6-methoxyquinoline | |
14860 | |
159.19 | |
Granular Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat,drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water f
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 226-077-2
missing translation for 'tsca' : TSCA
RUO – Research Use Only