missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxyquinoline, 98%

Product Code. 11479263
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479263 25 g 25g
11469263 5 g 5g
11489263 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479263 Supplier Thermo Scientific Alfa Aesar Supplier No. A11736.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5263-87-6 | C10H9NO | 159.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5263-87-6
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00006800
InChI Key HFDLDPJYCIEXJP-UHFFFAOYSA-N
Synonym quinoline, 6-methoxy, 6-methoxy-quinoline, methyl 6-quinolyl ether, p-quinanisole, unii-5s1u3125ad, chembl15200, 6-methoxy quinoline, pubchem5917, 6-methoxyquinoline, acmc-209l0q
PubChem CID 14860
ChEBI CHEBI:72822
IUPAC Name 6-methoxyquinoline
SMILES COC1=CC2=C(C=C1)N=CC=C2

Specifications

Melting Point 17°C to 20°C
Density 1.152
Boiling Point 141°C to 145°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.622
Quantity 25 g
Beilstein 115244
Sensitivity Light sensitive
Formula Weight 159.19
Percent Purity 98%
Chemical Name or Material 6-Methoxyquinoline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.