missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxyquinaldine, 97%, Thermo Scientific™

Product Code. 10573881 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
1g; Glass bottle
This item is not returnable. View return policy

Product Code. 10573881

Brand: Thermo Scientific Acros 146980010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1078-28-0
CAS Min % 96.0
CAS Max % 100.0
Melting Point 61°C to 65°C
Color Beige to Gray
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C11H11NO
MDL Number MFCD00006761
Quantity 1 g
Synonym 6-methoxyquinaldine, 2-methyl-6-methoxyquinoline, quinoline, 6-methoxy-2-methyl, quinaldine, 6-methoxy, 6-methoxy-2-methyl-quinoline, quinoline,6-methoxy-2-methyl, pubchem5910, acmc-2098xp, 2-methy-6-methoxyquinolinel
InChI Key NAGJQQFMJKMXJQ-UHFFFAOYSA-N
SMILES CC1=NC2=C(C=C1)C=C(C=C2)OC
IUPAC Name 6-methoxy-2-methylquinoline
Molecular Weight (g/mol) 173.21
PubChem CID 70648
Formula Weight 173.21
Percent Purity 97%
Physical Form Fluffy Crystalline Powder
Chemical Name or Material 6-Methoxyquinaldine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.