missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxypyridine-3-sulfonyl chloride, 97%

Product Code. 10155857 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10155857 5 g Glass bottle 5g
10564028 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10155857 Supplier Thermo Scientific Acros Supplier No. 429410050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 312300-42-8 | C6H6ClNO3S | 207.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312300-42-8
Molecular Formula C6H6ClNO3S
Molecular Weight (g/mol) 207.63
MDL Number MFCD06739108
InChI Key OMLIVJOTRYWHNY-UHFFFAOYSA-N
Synonym 6-methoxy-pyridine-3-sulfonyl chloride, 3-chlorosulphonyl-6-methoxypyridine, 6-methoxypyridin-3-ylsulfonyl chloride, 2-methoxypyridine-5-sulfonylchloride, 6-methoxy-3-pyridinesulfonyl chloride, 3-pyridinesulfonylchloride, 6-methoxy, 6-methoxypyridine-3-sulphonyl chloride, 5-chlorosulfonyl-2-methoxypyridine, 6-methoxypyridine-3-yl sulfonyl chloride
PubChem CID 22344709
IUPAC Name 6-methoxypyridine-3-sulfonyl chloride
SMILES COC1=NC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 42.0°C to 45.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 207.64
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 6-Methoxypyridine-3-sulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 6-Methoxypyridine-3-sulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.