missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methoxypyridine-2-carboxylic acid, 97%
Click to view available options
Quantity:
1 g
Unit Size:
1g
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Incompatible with
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Incompatible with
Specifications
Specifications
| CAS | 26893-73-2 |
| Melting Point | 130°C to 134°C |
| Molecular Formula | C7H7NO3 |
| MDL Number | MFCD06800962 |
| Quantity | 1 g |
| Synonym | 6-methoxypicolinic acid, 6-methoxy-2-pyridinecarboxylic acid, 2-methoxy-6-picolinic acid, 2-methoxy-6-picolinicacid, 2-pyridinecarboxylic acid, 6-methoxy, picolinic acid, 6-methoxy, 6-methoxy-pyridine-2-carboxylic acid, pubchem15165, 6-methoxypyridine-2-carboxylicacid, acmc-1cccg |
| Solubility Information | Slightly soluble in water. |
| InChI Key | KSWBODXXZITTPO-UHFFFAOYSA-N |
| SMILES | COC1=CC=CC(=N1)C(=O)O |
| IUPAC Name | 6-methoxypyridine-2-carboxylic acid |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction