missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxypyridine-2-boronic acid pinacol ester, 97%

Product Code. 11392289
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392289 1 g 1g
15497665 5 g 5g
15407675 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11392289 Supplier Thermo Scientific Alfa Aesar Supplier No. H27263.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1034297-69-2 | C12H18BNO3 | 235.09 g/mol

It is used as an active pharmaceutical intermediate and also in the design, structure-activity relationship, and characterization of the development candidate nvp-hsp990.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate and also in the design, structure-activity relationship, and characterization of the development candidate nvp-hsp990.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1034297-69-2
Molecular Formula C12H18BNO3
Molecular Weight (g/mol) 235.09
MDL Number MFCD06798266
InChI Key VYWCRVNLPOVYJH-UHFFFAOYSA-N
Synonym 2-methoxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxypyridine-2-boronic acid pinacol ester, pyridine,2-methoxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-methoxy pyridine-2-boronic acid pinacol ester, 6-methoxypyridine-2-boronicacidpinacolester, 6-methoxypyridin-2-ylboronic acid pinacol ester, 6-methoxypyridin-2-yl boronic acid pinacol ester, 2-methoxy-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-6-methoxy-2-pyridinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methyloxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
PubChem CID 17750508
IUPAC Name 2-methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=NC(=CC=C2)OC

Specifications

Melting Point 68°C to 72°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 235.09
Percent Purity 97%
Chemical Name or Material 6-Methoxypyridine-2-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.