missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxynaphthalene-2-carboxylic Acid (6-Methoxy-2-naphthoic Acid; 2-Carboxy-6-methoxynaphthalene), Mikromol™

Product Code. 17992756
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17992756

Brand: Mikromol MM0152.15

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

6-Methoxynaphthalene-2-carboxylic Acid (6-Methoxy-2-naphthoic Acid; 2-Carboxy-6-methoxynaphthalene)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Methoxynaphthalene-2-carboxylic Acid (6-Methoxy-2-naphthoic Acid; 2-Carboxy-6-methoxynaphthalene)
CAS 2471-70-7
Analyte or Component Names 6-Methoxynaphthalene-2-carboxylic Acid (6-Methoxy-2-naphthoic Acid; 2-Carboxy-6-methoxynaphthalene)
Shipping Condition Room Temperature
Molecular Formula C12 H10 O3
Certifications/Compliance ISO 17025
Synonym Naproxen Imp. O (EP), Naproxen USP Related Compound A, Naproxen USP RC A, 2-Carboxy-6-methoxynaphthalene, Naproxen Related Compound A, Naproxen Impurity O
InChI Formula InChI=1S/C12H10O3/c1-15-11-5-4-8-6-10(12(13)14)3-2-9(8)7-11/h2-7H,1H3,(H,13,14)
SMILES COc1ccc2cc(ccc2c1)C(=O)O
IUPAC Name 6-methoxynaphthalene-2-carboxylic acid
Molecular Weight (g/mol) 202.21
Quantity 100 mg
Formula Weight 202.063 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.