missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methoxynaphthalene-2-carboxylic Acid (6-Methoxy-2-naphthoic Acid; 2-Carboxy-6-methoxynaphthalene), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
6-Methoxynaphthalene-2-carboxylic Acid (6-Methoxy-2-naphthoic Acid; 2-Carboxy-6-methoxynaphthalene)
Specifications
Specifications
| Chemical Name or Material | 6-Methoxynaphthalene-2-carboxylic Acid (6-Methoxy-2-naphthoic Acid; 2-Carboxy-6-methoxynaphthalene) |
| CAS | 2471-70-7 |
| Analyte or Component Names | 6-Methoxynaphthalene-2-carboxylic Acid (6-Methoxy-2-naphthoic Acid; 2-Carboxy-6-methoxynaphthalene) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H10 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Naproxen Imp. O (EP), Naproxen USP Related Compound A, Naproxen USP RC A, 2-Carboxy-6-methoxynaphthalene, Naproxen Related Compound A, Naproxen Impurity O |
| InChI Formula | InChI=1S/C12H10O3/c1-15-11-5-4-8-6-10(12(13)14)3-2-9(8)7-11/h2-7H,1H3,(H,13,14) |
| SMILES | COc1ccc2cc(ccc2c1)C(=O)O |
| IUPAC Name | 6-methoxynaphthalene-2-carboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction