missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxyindole, 98%, Thermo Scientific™

Product Code. 10487592 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass Bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10487592 500 mg Glass Bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10487592 Supplier Thermo Scientific Acros Supplier No. 368145000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 3189-13-7
CAS Min % 97.5
CAS Max % 100.0
Melting Point 90.0°C to 94.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C9H9NO
MDL Number MFCD00022780
Quantity 500 mg
Beilstein 21, 22
Synonym 6-methoxyindole, 1h-indole, 6-methoxy, indole, 6-methoxy, 1h-indol-6-yl methyl ether, 6-methoxy indole, pubchem1701, acmc-209hph, opera_id_1903, 6-methyloxy-1h-indole
InChI Key QJRWYBIKLXNYLF-UHFFFAOYSA-N
SMILES COC1=CC=C2C=CNC2=C1
IUPAC Name 6-methoxy-1H-indole
Molecular Weight (g/mol) 147.18
PubChem CID 76659
Formula Weight 147.18
Percent Purity 98%
Physical Form Shiny Crystals
Chemical Name or Material 6-Methoxyindole, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.