missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxyindole, 98+%

Product Code. 11309635
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309635 1 g 1g
11319635 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309635 Supplier Thermo Scientific Alfa Aesar Supplier No. L19074.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3189-13-7 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3189-13-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00022780
InChI Key QJRWYBIKLXNYLF-UHFFFAOYSA-N
Synonym 6-methoxyindole, 1h-indole, 6-methoxy, indole, 6-methoxy, 1h-indol-6-yl methyl ether, 6-methoxy indole, pubchem1701, acmc-209hph, opera_id_1903, 6-methyloxy-1h-indole
PubChem CID 76659
IUPAC Name 6-methoxy-1H-indole
SMILES COC1=CC=C2C=CNC2=C1

Specifications

Melting Point 90°C to 94°C
Boiling Point 105°C (0.2 mmHg)
Quantity 1 g
Beilstein 116483
Sensitivity Light sensitive
Formula Weight 147.18
Percent Purity ≥98%
Chemical Name or Material 6-Methoxyindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.