missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-8-nitroquinoline, 99%, Thermo Scientific™

Product Code. 10173780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173780 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10173780 Supplier Thermo Scientific Acros Supplier No. 195600010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 85-81-4
CAS Min % 98.5
CAS Max % 100.0
Melting Point 158°C to 162°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H8N2O3
MDL Number MFCD00006802
Quantity 1 g
Beilstein 21, II, 54
Synonym quinoline, 6-methoxy-8-nitro, 6-methoxy-8-nitro-quinoline, 8-nitro-6-methoxyquinoline, methyl 8-nitro-6-quinolyl ether, unii-ats412o1xo, ats412o1xo, pubchem9433, acmc-209q7l, 5-21-03-00248 beilstein handbook reference, ksc655o7n
InChI Key MIMUSZHMZBJBPO-UHFFFAOYSA-N
SMILES COC1=CC(=C2C(=C1)C=CC=N2)[N+](=O)[O-]
IUPAC Name 6-methoxy-8-nitroquinoline
Molecular Weight (g/mol) 204.18
PubChem CID 6822
Formula Weight 204.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxy-8-nitroquinoline, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.