missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-naphthylglyoxal hydrate, Thermo Scientific™

Product Code. 10210100
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10210100

Brand: Thermo Scientific Maybridge MAY00154DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 745783-88-4
Packaging Amber Glass Bottle
Molecular Formula C13H10O3
MDL Number MFCD08533285
Quantity 5 g
Synonym 6-methoxy-2-naphthylglyoxal hydrate, 2-6-methoxynaphthalen-2-yl-2-oxoacetaldehyde hydrate, 6-methoxynaphthalen-2-yl oxo acetaldehyde-water 1/1, 2-naphthaleneacetaldehyde,6-methoxy-a-oxo-, hydrate 1:1, 2-2-methoxynaphthalen-6-yl-2-oxoacetaldehyde hydrate
InChI Key BAVQWNGBJSPFBJ-UHFFFAOYSA-N
SMILES COC1=CC2=CC=C(C=C2C=C1)C(=O)C=O
IUPAC Name 2-(6-methoxynaphthalen-2-yl)-2-oxoacetaldehyde;hydrate
Molecular Weight (g/mol) 214.22
PubChem CID 45074957
Formula Weight 214.22
Percent Purity 97%
Chemical Name or Material 6-Methoxy-2-naphthylglyoxal hydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.