missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-naphthylglyoxal hydrate, 98%, dry wt. basis

Product Code. 11369126
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369126 5 g 5g
11359126 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369126 Supplier Thermo Scientific Alfa Aesar Supplier No. L20260.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 745783-88-4 | C13H12O4 | 232.235 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 745783-88-4
Molecular Formula C13H12O4
Molecular Weight (g/mol) 232.235
MDL Number MFCD05664091
InChI Key CBFAHRRUWMTAQH-UHFFFAOYSA-N
Synonym 6-methoxynaphthylglyoxal hydrate, 2-6-methoxynaphthalen-1-yl-2-oxoacetaldehyde hydrate, 2-6-methoxy-1-naphthyl-2-oxo-acetaldehyde, pubchem12296, 2-6-methoxynaphthyl-2-oxoethanal, hydrate, 2-6-methoxy-1-naphthalenyl-2-oxoacetaldehyde hydrate, 6-methoxynaphthalen-1-yl oxo acetaldehyde-water 1/1, 2-6-methoxynaphthalen-1-yl-2-oxidanylidene-ethanal hydrate
PubChem CID 44118759
IUPAC Name 2-(6-methoxynaphthalen-1-yl)-2-oxoacetaldehyde;hydrate
SMILES COC1=CC2=C(C=C1)C(=CC=C2)C(=O)C=O.O

Specifications

Melting Point 125°C to 130°C
Quantity 5 g
Formula Weight 214.22 (Anhydrous)
Percent Purity 98%
Assay (dry wt.)
Chemical Name or Material 6-Methoxy-2-naphthylglyoxal hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.