missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-naphthylboronic Acid, TRC

Product Code. 30481859
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481859 250 mg 250mg
30480297 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481859 Supplier Toronto Research Chemicals Supplier No. TRCM264900250MG
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156641-98-4
Molecular Formula C11H11BO3
Molecular Weight (g/mol) 202.02
Synonym B-(6-Methoxy-2-naphthalenyl)boronic Acid, (6-Methoxy-2-naphthalenyl)boronic Acid, 6-Methoxynaphthalene-2-boronic Acid
IUPAC Name (6-methoxynaphthalen-2-yl)boronic acid
SMILES COc1ccc2cc(ccc2c1)B(O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H11BO3/c1-15-11-5-3-8-6-10(12(13)14)4-2-9(8)7-11/h2-7,13-14H,1H3
Formula Weight 202.0801
Chemical Name or Material 6-Methoxy-2-naphthylboronic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.