missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-naphthoic acid, MedChemExpress

Product Code. 16448846
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448846 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16448846 Supplier Medchem Express Supplier No. HYB2121
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 6-Methoxy-2-naphthoic acid is an NMDA receptor modulator extracted from patent WO 2012019106 A2.

6-Methoxy-2-naphthoic acid is an NMDA receptor modulator extracted from patent WO 2012019106 A2.

Specifications

Chemical Name or Material 6-Methoxy-2-naphthoic acid
CAS 2471-70-7
Color White
Purity Grade Notes Research
Molecular Formula C12H10O3
Quantity 100 mg
Synonym Naproxen impurity O
Solubility Information DMSO : 60 mg/mL (296.72 mM; Need ultrasonic and warming)
SMILES O=C(C1=CC=C2C=C(OC)C=CC2=C1)O
Molecular Weight (g/mol) 202.21
Formula Weight 202.21
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.