missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-methylbenzothiazole, 97%, Thermo Scientific Chemicals

Catalog Number 15425147
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2941-72-2
C9H9NOS
179.237
MFCD00005795
DYHLJSUORLPGNT-UHFFFAOYSA-N
6-methoxy-2-methylbenzo d thiazole, 6-methoxy-2-methylbenzothiazole, benzothiazole, 6-methoxy-2-methyl, 2-methyl-6-methoxybenzothiazole, 6-methoxy-2-methyl-benzothiazole, benzothiazole,6-methoxy-2-methyl, 6-methoxy-2-methylbenzothiazol, #, benzothiazole, 6-methoxy-2-methyl-6ci,7ci,8ci,9ci
76254
6-methoxy-2-methyl-1,3-benzothiazole
CC1=NC2=C(S1)C=C(C=C2)OC
This item is not returnable. View return policy
2941-72-2
C9H9NOS
179.237
MFCD00005795
DYHLJSUORLPGNT-UHFFFAOYSA-N
6-methoxy-2-methylbenzo d thiazole, 6-methoxy-2-methylbenzothiazole, benzothiazole, 6-methoxy-2-methyl, 2-methyl-6-methoxybenzothiazole, 6-methoxy-2-methyl-benzothiazole, benzothiazole,6-methoxy-2-methyl, 6-methoxy-2-methylbenzothiazol, #, benzothiazole, 6-methoxy-2-methyl-6ci,7ci,8ci,9ci
76254
6-methoxy-2-methyl-1,3-benzothiazole
CC1=NC2=C(S1)C=C(C=C2)OC

6-Methoxy-2-methylbenzothiazole was used as an inhibitor of aryl hydrocarbon hydroxylase (PB/AHH) and aminopyrine N-demethylase (ADPM) in hepatic microsomes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methoxy-2-methylbenzothiazole was used as an inhibitor of aryl hydrocarbon hydroxylase (PB/AHH) and aminopyrine N-demethylase (ADPM) in hepatic microsomes.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.204
Flash Point 109°C (228°F)
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 179.24
Percent Purity 97%
Chemical Name or Material 6-Methoxy-2-methylbenzothiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'