missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Mercaptopurine monohydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
6-Mercaptopurine is a widely used antileukemic agent that inhibits de novopurine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA. It is also an antitumor agent and induces apoptosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Mercaptopurine is a widely used antileukemic agent that inhibits de novopurine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA. It is also an antitumor agent and induces apoptosis.
Solubility
Soluble in dilute solutions of alkali hydroxides, hot alcohol, and 1 N NaOH (50 mg/ml). Insoluble in water.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
6-Mercaptopurine is a widely used antileukemic agent that inhibits de novopurine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA. It is also an antitumor agent and induces apoptosis.
Solubility
Soluble in dilute solutions of alkali hydroxides, hot alcohol, and 1 N NaOH (50 mg/ml). Insoluble in water.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
Specifications
Melting Point | ∼315°C (decomposition) |
Quantity | 25 g |
Beilstein | 4012091 |
Merck Index | 14,5871 |
Solubility Information | Soluble in dilute solutions of alkali hydroxides,hot alcohol,and 1 N NaOH (50mg/ml). Insoluble in water. |
Formula Weight | 170.19 (152.17 Anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | 6-Mercaptopurine monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'