missing translation for 'onlineSavingsMsg'
Learn More

6-Mercaptopurine monohydrate, 98%

Product Code. 11484853
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484853 5 g 5g
11474853 1 g 1g
11494853 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484853 Supplier Thermo Scientific Alfa Aesar Supplier No. A12197.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6112-76-1 | C5H6N4OS | 170.19 g/mol

6-Mercaptopurine is a widely used antileukemic agent that inhibits de novopurine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA. It is also an antitumor agent and induces apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Mercaptopurine is a widely used antileukemic agent that inhibits de novopurine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA. It is also an antitumor agent and induces apoptosis.

Solubility
Soluble in dilute solutions of alkali hydroxides, hot alcohol, and 1 N NaOH (50 mg/ml). Insoluble in water.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6112-76-1
Molecular Formula C5H6N4OS
Molecular Weight (g/mol) 170.19
MDL Number MFCD03854445,MFCD01461928
InChI Key WFFQYWAAEWLHJC-UHFFFAOYSA-N
Synonym 6-mercaptopurine monohydrate, 6-mercaptopurine hydrate, mercaptopurine monohydrate, mercaptopurine hydrate, 6h-purine-6-thione, 1,7-dihydro-, monohydrate, 6,7-dihydro-3h-purine-6-thione hydrate, purine-6-thiol monohydrate, 1,7-dihydro-6h-purine-6-thione monohydrate, purine-6-thiol, monohydrate, unii-e7wed276i5
PubChem CID 2724350
ChEBI CHEBI:31822
IUPAC Name 3,7-dihydropurine-6-thione;hydrate
SMILES O.S=C1N=CNC2=C1NC=N2

Specifications

Melting Point ∼315°C (decomposition)
Quantity 5 g
Beilstein 4012091
Merck Index 14,5871
Solubility Information Soluble in dilute solutions of alkali hydroxides,hot alcohol,and 1 N NaOH (50mg/ml). Insoluble in water.
Formula Weight 170.19 (152.17 Anhydrous)
Percent Purity 98%
Chemical Name or Material 6-Mercaptopurine monohydrate
compliance-icons
Product Identifier
  • 6-Mercaptopurine monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 6-Mercaptopurine monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.