missing translation for 'onlineSavingsMsg'
Learn More

6-Mercaptopurine, MedChemExpress

Product Code. 16451957
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16451957

Brand: Medchem Express HY13677

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 6-Mercaptopurine is a purine analogue which acts as an antagonist of the endogenous purines and has been widely used as antileukemic agent and immunosuppressive drug.

6-Mercaptopurine is a purine analogue which acts as an antagonist of the endogenous purines and has been widely used as antileukemic agent and immunosuppressive drug.

Specifications

Chemical Name or Material 6-Mercaptopurine
Physical Form Solid
CAS 50-44-2
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C5H4N4S
For Use With (Application) Cancer-programmed cell death
Synonym Mercaptopurine 6-MP
Solubility Information DMSO : 35.71 mg/mL (234.66 mM; Need ultrasonic)
SMILES S=C1NC=NC2=C1NC=N2
Molecular Weight (g/mol) 152.18
Formula Weight 152.18
Percent Purity 96.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.