missing translation for 'onlineSavingsMsg'
Learn More

6-Maleimidohexanoic acid

Product Code. 15462677
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15462677

Brand: Thermo Scientific Alfa Aesar 046384.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55750-53-3 | C10H13NO4 | 211.217 g/mol

6-Maleimidohexanoic acid is used as a probe for introducing maleimides groups into biomolecules and active pharmaceutical ingredients. Further, it is used with N-hydroxysuccinimide ester as a bifunctional cross-linking reagent. In addition to this, it acts as a probe for thiol groups in membrane proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55750-53-3
Molecular Formula C10H13NO4
Molecular Weight (g/mol) 211.217
MDL Number MFCD00043140
InChI Key WOJKKJKETHYEAC-UHFFFAOYSA-N
Synonym 6-maleimidocaproic acid, 6-maleimidohexanoic acid, 6-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl hexanoic acid, 6-maleimidocaproicacid, 1h-pyrrole-1-hexanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 5-cooh, n-maleoyl-6-aminocaproic acid, n-maleoyl-6-aminohexanoic acid, 1h-pyrrole-1-hexanoicacid, 2,5-dihydro-2,5-dioxo, 6-2,5-dioxopyrrol-1-yl hexanoic acid
PubChem CID 573683
IUPAC Name 6-(2,5-dioxopyrrol-1-yl)hexanoic acid
SMILES C1=CC(=O)N(C1=O)CCCCCC(=O)O

Specifications

Color White to Yellow
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water,methanol,ethanol,dimethyl sulfoxide. Insoluble in ether.
Formula Weight 211.22
Physical Form Powder
Chemical Name or Material 6-Maleimidohexanoic acid
compliance-icons
Product Identifier
  • 6-Maleimidohexanoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Zinc oxalate dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.