missing translation for 'onlineSavingsMsg'
Learn More

6-Maleimidohexanoic acid N-hydroxysuccinimide ester, MedChemExpress

Product Code. 16457966
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16457966 100 mg 100mg
16454127 50 mg 50mg
16401966 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16457966 Supplier Medchem Express Supplier No. HY78961
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 6-Maleimidohexanoic acid N-hydroxysuccinimide ester (EMCS) is a heterobifunctional cross-linking reagent. EMCS is used as a unique and useful reagent for preparation of hapten conjugate and enzyme immunoconjugates.

6-Maleimidohexanoic acid N-hydroxysuccinimide ester (EMCS) is a heterobifunctional cross-linking reagent. EMCS is used as a unique and useful reagent for preparation of hapten conjugate and enzyme immunoconjugates.

Specifications

Chemical Name or Material 6-Maleimidohexanoic acid N-hydroxysuccinimide ester
CAS 55750-63-5
Color White
Purity Grade Notes Research
Molecular Formula C14H16N2O6
Quantity 100 mg
Synonym EMCS
Solubility Information DMSO : 230 mg/mL (746.05 mM; Need ultrasonic)
SMILES O=C(C=C1)N(CCCCCC(ON2C(CCC2=O)=O)=O)C1=O
Molecular Weight (g/mol) 308.29
Formula Weight 308.29
Percent Purity 95.75%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.