missing translation for 'onlineSavingsMsg'
Learn More

6-Maleimidocaproic acid N-succinimidyl ester, 95%

Product Code. 15653694 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
15653694 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15653694 Supplier Thermo Scientific Supplier No. H6599603
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 55750-63-5 | C14H16N2O6 | 308.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55750-63-5
Molecular Formula C14H16N2O6
Molecular Weight (g/mol) 308.29
MDL Number MFCD00043043
InChI Key VLARLSIGSPVYHX-UHFFFAOYSA-N
Synonym n-succinimidyl 6-maleimidohexanoate, emcs, 6-maleimidohexanoic acid n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 6-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl hexanoate, n-succinimidyl 6-maleimidocaproate, n-6-maleimidocaproyloxy succinimide, 2,5-dioxopyrrolidin-1-yl 6-2,5-dioxopyrrol-1-yl hexanoate, nmicos, 1h-pyrrole-2,5-dione, 1-6-2,5-dioxo-1-pyrrolidinyl oxy-6-oxohexyl
PubChem CID 5091655
IUPAC Name 2,5-dioxopyrrolidin-1-yl 6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoate
SMILES O=C(CCCCCN1C(=O)C=CC1=O)ON1C(=O)CCC1=O

Specifications

Melting Point 69°C to 74°C
Color White to Brown
Quantity 1 g
Beilstein 1499815
Sensitivity Light and moisture sensitive; Hygroscopic
Formula Weight 308.29
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 6-Maleimidocaproic acid N-succinimidyl ester
compliance-icons
Product Identifier
  • 6-Maleimidocaproic acid N-succinimidyl ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 6-Maleimidocaproic acid N-succinimidyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.