missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Keto Fulvestrant, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Keto Fulvestrant |
| Analyte or Component Names | 6-Keto Fulvestrant |
| Shipping Condition | Room Temperature |
| Molecular Formula | C32H45F5O4S |
| Synonym | (7α,17β)-7-[9-[4,4,5,5,5-Pentafluoropentyl)sulfinyl]nonyl]-3,17β-dihydroxyestra-1,3,5(10)-trien-6-one, Fulvestrant Impurity F |
| InChI Formula | InChI=1S/C32H45F5O4S/c1-30-17-15-23-22-12-11-21(38)20-25(22)29(40)24(28(23)26(30)13-14-27(30)39)10-7-5-3-2-4-6-8-18-42(41)19-9-16-31(33,34)32(35,36)37/h11-12,20,23-24,26-28,38-39H,2-10,13-19H2,1H3/t23-,24+,26+,27+,28+,30+,42?/m1/s1 |
| Solution Type | Neat |
| SMILES | OC1=CC(C([C@@H](CCCCCCCCCS(CCCC(F)(F)C(F)(F)F)=O)[C@]2([H])[C@]3([H])CC[C@@]4(C)[C@@]2([H])CC[C@@H]4O)=O)=C3C=C1 |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 620.75 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?