missing translation for 'onlineSavingsMsg'
Learn More

6-Isocyanatochroman, 97%, Thermo Scientific™

Product Code. 10031534
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10031534

Brand: Thermo Scientific Maybridge CC32806DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1002727-87-8
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD12198109
InChI Key YNAWMZLOWDUDQF-UHFFFAOYSA-N
Synonym 6-isocyanatochroman, 6-isocyanato-3,4-dihydro-2h-1-benzopyran, chroman-6-isocyanate
PubChem CID 43811033
IUPAC Name 6-isocyanato-3,4-dihydro-2H-chromene
SMILES C1CC2=C(C=CC(=C2)N=C=O)OC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material 6-Isocyanatochroman
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.